Learn More
Analytical Sales & Services Lancer C18 Prep Column. 120Â, 5µm, 10cm x 20mm ID
Supplier:  Analytical Sales & Services ADV531020
Encompass
Lancer C18 Prep Column. 120Â, 5µm, 10cm x 20mm ID
Catalog No. 50-008-05448