Learn More
Getinge USA LancerClean Detergent 2 x 2.5 Gallon
Supplier:  Getinge USA LCSLCD2.5S
Encompass
LancerClean Detergent is available in three solutions (LCD-P , LCD-S , LCD-H) that are capable of cleaning all different types of contaminants including laboratory soils, organic residues, petroleum applications and other hard to clean soils.The laboratory detergent is specifically formulated for many types of goods including glassware, surgical instruments and many other labware items.* Made to order, non-returnable. Can not ship air.*
Catalog No. NC2941064