Learn More
Medchemexpress LLC Landiolol hydrochloride | 144481-98-1 | MFCD01937430 | >=98.0% | 546.05 g/mol | C25H40ClN3O8 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100607A10MM 1ML
Encompass_Preferred
Landiolol hydrochloride is a highly selective, ultra-short-acting β1-adrenergic receptor antagonist formulated for research use. It is suitable for in vitro pharmacology and biochemical studies, and is offered as both a ready-to-use solution and as solid material for analytical and formulation purposes.
  • Highly selective β1-adrenergic receptor antagonist.
  • Ultra-short-acting pharmacological profile suitable for acute studies.
  • Available as a 10 mM solution in DMSO and as solid forms for flexibility.
  • Provided with molecular and analytical characterization data.
  • Designed for in vitro receptor pharmacology and biochemical assays.
Catalog No. 50-002-14639