Learn More
TARGETMOL CHEMICALS INC Landiolol hydrochloride 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T502850MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Landiolol hydrochloride (ONO-1101 hydrochloride) is an ultra-short-acting beta1 selective adrenoceptor antagonist. Purity 100%
Catalog No. 50-287-7335