Learn More
TARGETMOL CHEMICALS INC Lanepitant 2HCl 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T256161MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Lanepitant 2HCl is a non-peptide neurokinin-1 receptor antagonist with analgesic and anti-inflammatory activity.Lanepitant 2HCl inhibits the binding of Substance P to the neurokinin-1 receptor, inhibits neurogenic inflammation and pain transmission, and can be used in the study of painful neuropathic disorders, such as migraines. Purity 98.89%
Catalog No. 50-287-6489