Learn More
Medchemexpress LLC Lanicemine dihydrochloride | 153322-06-6 | 99.9% | 271.19 g/mol | C13H16Cl2N2 | 1 ML

Supplier: Medchemexpress LLC HY108235A10MM 1ML
Lanicemine dihydrochloride is a low-trapping NMDA receptor channel blocker (AZD6765) provided as a 10 mM solution in DMSO for laboratory research. It is used in pharmacology and neuroscience studies to examine NMDA receptor modulation and antidepressant-like effects. The compound has CAS 153322-06-6 and a molecular weight of 271.19 g/mol.
- Supplied as 10 mM solution in DMSO, 1 mL
- High stated purity (99.94%) suitable for research applications
- Low-trapping NMDA receptor antagonist for pharmacological studies
- Relevant for studies of antidepressant mechanisms and synaptic transmission
- Molecular formula C13H16Cl2N2; molecular weight 271.19 g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.