Learn More
Medchemexpress LLC Laninamivir | 203120-17-6 | 346.34 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14818100MG
Encompass_Preferred
Laninamivir is a potent influenza neuraminidase (NA) inhibitor. It effectively inhibits common oseltamivir-resistant viruses, including those with the His274Tyr substitution, and demonstrates potency against various influenza strains.
  • Potent influenza neuraminidase (NA) inhibitor
  • Inhibits common oseltamivir-resistant viruses
  • Effective against avian H12N5 NA (N5), pH1N1 N1 NA (p09N1), and A/RI/5+/1957 H2N2 N2 (p57N2)
  • Exhibits a similar binding mode to Zanamivir
  • Suitable for laboratory chemical applications and substance manufacturing
Catalog No. 50-003-62845