Learn More
Medchemexpress LLC Laninamivir | 203120-17-6 | 99.97% | 346.34 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1481850MG
Encompass_Preferred
Laninamivir is a potent influenza neuraminidase (NA) inhibitor. It is effective against common oseltamivir-resistant viruses, including those with the His274Tyr substitution, and shows a similar binding mode to Zanamivir. This product is for research use only.
  • Potent influenza neuraminidase (NA) inhibitor
  • Effective against common oseltamivir-resistant viruses
  • Similar binding mode to Zanamivir
  • IC50s of 0.90 nM for avian H12N5 NA (N5), 1.83 nM for pH1N1 N1 NA (p09N1), and 3.12 nM for A/RI/5+/1957 H2N2 N2 (p57N2)
Catalog No. 50-003-62241