Learn More
Selleck Chemical LLC Laninamivir E4879-1MG
SDP
Supplier:  Selleck Chemical LLC E48791MG
Encompass_Preferred
Laninamivir(R-125489) is a potent inhibitor of influenza neuraminidase(NA)with IC50s of 0 90 nM 1 83 nM and 3 12 nM for avian H12N5 NA (N5) pH1N1 N1 NA (p09N1) and A/RI/5 /1957 H2N2 N2 (p57N2) respectively It exhibits long-acting anti-influenza virus activity
Catalog No. 50-000-41847