Learn More
Medchemexpress LLC Lanosta-7,9(11),24-trien-3-one, 26-hydroxy-, (24E)- | 104700-97-2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN39255MG
Encompass_Preferred
Ganoderol A is a terpenoid extracted from Ganoderma lucidum with antimicrobial activities. It inhibits the cholesterol synthesis pathway and exhibits significant anti-inflammatory activity and protection against ultraviolet A (UVA) damage. This product is intended for research use only.
  • Antimicrobial activities
  • Inhibits cholesterol synthesis pathway
  • Anti-inflammatory activity
  • Protection against ultraviolet A (UVA) damage
  • Solid appearance
  • White to off-white color
  • Structure classification: terpenoids, triterpenes
  • Initial source: plants, Polyporaceae
  • Purity: 97.0%
Catalog No. 50-004-18305