Learn More
Selleck Chemical LLC Lanraplenib S9715-5MG
SDP
Supplier:  Selleck Chemical LLC S97155MG
Encompass_Preferred
Lanraplenib (GS-SYK)is a potent highly selective and orally active inhibitor of Spleen Tyrosine Kinase (SYK) with IC50 of 9 5 nM Lanraplenib inhibits SYK activity in platelets via the glycoprotein VI (GPVI) receptor without prolonging bleeding time (BT) in monkeys or humans
Catalog No. 50-000-24797