Learn More
Medchemexpress LLC HY-109091B 5mg Medchemexpress, Lanraplenib (succinate) CAS:1800047-00-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109091B5MG
Encompass_Preferred
Medchemexpress, HY-109091B 5mg Lanraplenib (succinate) CAS:1800047-00-0 Lanraplenib succinate (GS-9876 succinate) is a highly selective and orally active SYK inhibitor (IC50=9.5 nM) in development for the treatment of inflammatory diseases. Lanraplenib succinate (GS-9876 succinate) inhibits SYK activity in platelets via the glycoprotein VI (GPVI) receptor without prolonging bleeding time (BT) in monkeys or humans[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8674