Learn More
Selleck Chemical LLC Lanreotide acetate P1088-25MG
SDP
Supplier:  Selleck Chemical LLC P108825MG
Encompass_Preferred
Lanreotide (Laromustine Angiopeptin BIM 23014 Dermopeptin Ipstyl ITM-014 Somatulina Somatuline Lanreotidum) acetate an analog of the native somatostatin peptide is a physiological inhibitor of growth hormone (GH) Lanreotide also has antitumoral effects
Catalog No. 50-000-40883