Learn More
Medchemexpress LLC Lanreotide (acetate) | 2378114-72-6 | 99.8% | 1156.38 | 5 MG
Supplier: Medchemexpress LLC 5MGHYP1959A
Lanreotide acetate is a somatostatin analogue with antineoplastic activity, primarily used in research for conditions such as carcinoid syndrome.
- Enhances radiation-induced apoptosis
- Inhibits GH-secreting pituitary adenoma cell proliferation and hormone release in vitro
- Induces tumor growth inhibition in male nude mice