Learn More
Apexbio Technology LLC Lansoprazole 103577-45-3 1g
SDP
Supplier:  Apexbio Technology LLC A12291000
Encompass_Preferred
Lansoprazole (103577-45-3) is a small-molecule inhibitor targeting gastric H K -ATPase an enzyme responsible for gastric acid secretion via proton pump activity It is designed to inhibit H K -ATPase activity thereby suppressing gastric acid production Lansoprazole exerts its biological activity primarily through selective inhibition of the H K -ATPase enzyme In experimental models lansoprazole demonstrates inhibitory activity with an IC50 value of approximately 6 3 M for enzyme activity and 0 09 M for suppression of histamine-stimulated gastric acid secretion Based on these pharmacological properties lansoprazole holds research potential in studies of gastric acid-related pathophysiology gastroprotective mechanisms and ulcer formation processes
Catalog No. 50-000-93116