Learn More
Medchemexpress LLC HY-13662 500mg , Lansoprazole CAS:103577-45-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13662500MG
Encompass_Preferred
HY-13662 500mg Lansoprazole CAS: 103577-45-3 Lansoprazole (AG 1749) is an orally active proton pump inhibitor which prevents the stomach from producing acid. Lansoprazole (AG 1749) is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0794