Learn More
Strem, An Ascensus Company CAS# 7439-91-0. 25g. Lanthanum chips (99.6% REO). MFCD00011066
SDP
Supplier:  Strem, An Ascensus Company 93574625G
Encompass
CAS# 7439-91-0. 25g. Lanthanum chips (99.6% REO). MFCD00011066. Molecular Weight: 138.90. Molecular Formula: La. Color/form: (packed in mineral oil) Strem# 93-5746. http://www.strem.com/catalog/v/93-5746/
Catalog No. 50-901-15862