Learn More
Strem, An Ascensus Company CAS# 100587-90-4. 100g. Lanthanum(III) acetate hydrate (99.9%-La) (REO). MFCD00150118
SDP
Supplier:  Strem, An Ascensus Company 935701100G
Encompass
CAS# 100587-90-4. 100g. Lanthanum(III) acetate hydrate (99.9%-La) (REO). MFCD00150118. Molecular Weight: 316.05. Molecular Formula: La(OOCCH3)3∙XH2O. Color/form: white xtl. Strem# 93-5701. http://www.strem.com/catalog/v/93-5701/
Catalog No. 50-901-15823