Learn More
Strem, An Ascensus Company CAS# 1312-81-8. 500g. Lanthanum(III) oxide (99.99%-La) (REO) PURATREM. MFCD00011071
SDP
Supplier:  Strem, An Ascensus Company 935716500G
Encompass
CAS# 1312-81-8. 500g. Lanthanum(III) oxide (99.99%-La) (REO) PURATREM. MFCD00011071. Molecular Weight: 325.82. Molecular Formula: La2O3. Color/form: white pwdr. Strem# 93-5716. http://www.strem.com/catalog/v/93-5716/
Catalog No. 50-901-15840