Learn More
Medchemexpress LLC Lanuginosine | 23740-25-2 | 98.3% | 5 MG

Supplier: Medchemexpress LLC HY1193985MG
Lanuginosine is an aporphine alkaloid research compound with reported biological activities including apoptosis induction, acetylcholinesterase (AChE) inhibition (IC50: 10.9 μM), and inhibition of Aβ aggregation. It is used in anticancer and Alzheimer's disease research.
- Alkaloid research compound with formula C18H11NO4.
- Molecular weight 305.28 g·mol⁻1.
- Purity 98.3% (as listed).
- Reported activities include apoptosis induction, AChE inhibition (IC50: 10.9 μM), and inhibition of Aβ aggregation.
- Supplied as a 5 mg pack for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.