Learn More
Apexbio Technology LLC LAP 85073-19-4 500mg
SDP
Supplier:  Apexbio Technology LLC BA9550500
Encompass_Preferred
Lithium phenyl-2 4 6-trimethylbenzoylphosphinate (LAP CAS 85073-19-4) is a photoinitiator that generates free radicals upon exposure to light Through this radical formation LAP facilitates photopolymerization reactions commonly employed in biomedical research such as the fabrication of hydrogels for 3D bioprinting applications The free radicals produced by LAP can exert cytotoxic and potentially mutagenic effects under experimental conditions necessitating careful optimization in cell-based assays LAP is widely utilized for controlled light-mediated crosslinking in tissue engineering and biofabrication studies
Catalog No. 50-000-87918