Learn More
Medchemexpress LLC Lapachol | 84-79-7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN6961100MG
Encompass_Preferred
Lapachol is a natural naphthoquinone known for its diverse biological activities. It functions as an orally active and potent DHODH inhibitor, demonstrating immunosuppressive effects on lymphocytes by blocking DHODH activity and inhibiting pyrimidine synthesis. Furthermore, Lapachol acts as a vitamin K antagonist, exhibiting antitumor activity by inhibiting DNA and RNA synthesis in neoplastic cells, and also possesses anti-Leishmania properties.
  • Lapachol is a natural naphthoquinone.
  • It is an orally active, potent DHODH inhibitor.
  • Lapachol exhibits immunosuppressive activity on lymphocytes by blocking DHODH activity and inhibiting pyrimidine synthesis.
  • It acts as a vitamin K antagonist with antitumor activity.
  • Lapachol can inhibit DNA and RNA synthesis in neoplastic cells.
  • It possesses anti-Leishmania activity.
Catalog No. 50-004-20024