Learn More
Medchemexpress LLC Lapaquistat | 189059-71-0 | 99.8% | 603.10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY149255MG
Encompass_Preferred
Lapaquistat is a cholesterol biosynthesis inhibitor and the active metabolite of Lapaquistat acetate. It can decrease statin-induced myotoxicity in lipid-lowering therapy.
  • Inhibits cholesterol biosynthesis in differentiated RD cells with an IC50 of 36 nM.
  • Potently inhibits cholesterol synthesis in RD cells, with an IC25 exceeding 100 μM.
  • Concentration-dependently inhibits cholesterol biosynthesis in human skeletal myocytes, with an IC50 of 45 nM.
  • For research use only.
Catalog No. 50-003-59357