Learn More
Medchemexpress LLC Lapaquistat | 189059-71-0 | 99.81% | 603.10 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY149251MG
Encompass_Preferred
Lapaquistat is a cholesterol biosynthesis inhibitor and the active metabolite of Lapaquistat acetate. It can decrease statin-induced myotoxicity in lipid-lowering therapy. This product is suitable for laboratory research in cholesterol biosynthesis inhibition.
  • Inhibits cholesterol biosynthesis in differentiated RD cells with an IC50 of 36 nM
  • Potently inhibits cholesterol synthesis in RD cells
  • Concentration-dependently inhibits cholesterol biosynthesis in human skeletal myocytes with an IC50 of 45 nM
Catalog No. 50-003-59322