Learn More
Medchemexpress LLC Lapaquistat (T-91485) | 189059-71-0 | 99.8% | 603.10 | C31H39ClN2O8 | 10 MG

Supplier: Medchemexpress LLC HY1492510MG
Lapaquistat (T-91485) is a cholesterol biosynthesis inhibitor and the active metabolite of lapaquistat acetate. It is supplied as a research-grade solid for laboratory use in studies of lipid metabolism and for investigating approaches to reduce statin-induced myotoxicity. Analytical characterization and storage guidance accompany the material.
- High purity, typically ~99.8% by HPLC.
- Molecular weight 603.10 g/mol.
- Chemical formula C31H39ClN2O8.
- CAS number 189059-71-0.
- Supplied as a powder; recommended storage -20°C for long-term stability.
- Suitable for in vitro and in vivo research applications in lipid metabolism.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.