Learn More
Medchemexpress LLC Lapatinib | 231277-92-2 | 99.8% | C29H26ClFN4O4S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY50898100MG
Encompass_Preferred
Lapatinib (GW572016) is a potent, orally active inhibitor of the ErbB-2 and EGFR tyrosine kinase domains. It has IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively. Lapatinib treatment inhibits receptor autophosphorylation of EGFR and ErbB-2 in a dose-responsive manner, and the phosphorylation of serine 473 of AKT was also inhibited in a dose-dependent manner. It also shows selective inhibition of the proliferation of human tumor cell lines and induces G1 arrest. In vivo, Lapatinib inhibits tumor xenograft growth in a dose-responsive manner.
  • Potent, orally active inhibitor of ErbB-2 and EGFR tyrosine kinase domains.
  • Inhibits receptor autophosphorylation of EGFR and ErbB-2.
  • Inhibits phosphorylation of serine 473 of AKT.
  • Shows selective inhibition of human tumor cell line proliferation.
  • Induces G1 arrest.
  • Inhibits tumor xenograft growth in vivo.
Catalog No. 50-003-58158