Learn More
Selleck Chemical LLC Lapatinib 25mg 231277-92-2 GW-572016, GSK572016
SDP
Supplier:  Selleck Chemical LLC S211125MG
Encompass_Preferred
Lapatinib (GSK572016), used in the form of Lapatinib Ditosylate, is a potent EGFR and ErbB2 inhibitor with IC50 of 10.8 and 9.2 nM in cell-free assays, respectively. Lapatinib induces ferroptosis and autophagic cell death. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2063