Learn More
Selleck Chemical LLC Lapatinib S2111-1g
SDP
Supplier:  Selleck Chemical LLC S21111G
Encompass_Preferred
Lapatinib is a potent EGFR and ErbB2 inhibitor with IC50 of 10 8 and 9 2 nM in cell-free assays respectively Lapatinib induces ferroptosis and autophagic cell death
Catalog No. 50-310-9401