Learn More
TARGETMOL CHEMICALS INC Lapatinib 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0078500MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Lapatinib (GW572016) is an inhibitor of ErbB2 and EGFR (IC50=9.2/10.8 nM) with oral activity. Lapatinib has antitumor activity and can be used to treat advanced or metastatic breast cancer with HER2 overexpression. Purity 99.89%
Catalog No. 50-288-0680