Learn More
Medchemexpress LLC HY-50898 1g , Lapatinib GW572016 CAS:231277-92-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY508981G
Encompass_Preferred
Medchemexpress, HY-50898 1g Lapatinib GW572016 CAS:231277-92-2 Formula:C29H26ClFN4O4S IC50: 10.2 nM (EGFR), 9.8 nM (ErbB2) Purity:98% Lapatinib is a potent EGFR and ErbB2 inhibitor with IC 50 of 10.2 and 9.8 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2278