Learn More
Cayman Chemical LapatInIb-d4tosylate 1mg
SDP
Supplier:  Cayman Chemical 254591 MG
Encompass_Preferred
An internal standard for the quantification of lapatinib by GC- or LC-MS
Catalog No. 50-293-6257