Learn More
TARGETMOL CHEMICALS INC LAPATINIB DITOSYLATE 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6235500MG
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 200 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lapatinib Ditosylate (Tykerb ditosylate) is an effective EGFR and ErbB2 inhibitor (IC50 10.8/9.2 nM for EGFR/ErbB2). purity: 99%
Catalog No. 50-257-6562