Learn More
Selleck Chemical LLC Lapatinib Ditosylate S1028-1g
SDP
Supplier:  Selleck Chemical LLC S10281G
Encompass_Preferred
Lapatinib Ditosylate is a potent EGFR and ErbB2 inhibitor with IC50 of 10 8 and 9 2 nM in cell-free assays respectively
Catalog No. 50-310-8405