Learn More
Medchemexpress LLC Lapatinib ditosylate monohydrate | 388082-78-8 | 100.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY50898B50MG
Encompass_Preferred
Lapatinib ditosylate monohydrate is a potent, orally active inhibitor of the ErbB-2 and EGFR tyrosine kinase domains. It has demonstrated IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively. This compound is for research use only.
  • Potent, orally active inhibitor of ErbB-2 and EGFR tyrosine kinase domains.
  • Inhibits receptor autophosphorylation of EGFR and ErbB-2.
  • Selectively inhibits proliferation of human tumor cell lines overexpressing EGFR or ErbB-2.
  • Induces G1 arrest in HN5 cells.
  • Inhibits tumor xenograft growth in vivo.
  • Associated with numerous clinical trials for various cancers.
Catalog No. 50-003-59082