Learn More
Selleck Chemical LLC Laprituximab emtansine-1MG
SDP
Supplier:  Selleck Chemical LLC D40371MG
Encompass_Preferred
Laprituximab emtansine (IMGN-289) is an anti-EGFR (epidermal growth factor receptor receptor tyrosine-protein kinase erbB-1 ERBB1 HER1 HER-1 ERBB) chimeric monoclonal antibody conjugated to maytansinoid DM1 via a succinimidyl-4-(N-maleimidomethyl) cyclohexane-1-carboxylate (SMCC) linker forming a nonreducible thioether bond
Catalog No. 50-313-4613