Learn More
Cayman Chemical LAPtrIfluoroacetate salt 10mg
SDP
Supplier:  Cayman Chemical 3698210 MG
Encompass_Preferred
An antiviral peptide; inhibits the HCV 5’-UTR-PTB and HCV 5’-UTR-PCBP2 interaction; inhibits HCV IRES-mediated translation in a reporter assay using Huh7 cells at 60 µM
Catalog No. 50-291-4999