Learn More
Medchemexpress LLC HY-106268A 10mg Medchemexpress, Larazotide (acetate) CAS:881851-50-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106268A10MG
Encompass_Preferred
Medchemexpress, HY-106268A 10mg Larazotide (acetate) CAS:881851-50-9 Larazotide acetate is a synthetic peptide. Larazotide acetate acts as a tight junction regulator and reverses leaky junctions to their normally closed state. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8775