Learn More
Genesee Scientific Corporation LARGE ICE PAN,YELLOW 9L, W/LID
SDP
Supplier:  Genesee Scientific Corporation 18310Y
Encompass
Genesee Scientific 18-310Y, Large Ice Pan with Lid, Yellow 9 Liters, 1 Ice Pan/Unit9 Liters1 Ice Pan/UnitBrand: Genesee ScientificExcellent insulation, stackable, lightweight and leak-proof designSuperior durability - no sweat, no rust, no deformation and virtually unbreakableChemical resistance - can withstand dry ice, chemical solvents and liquid nitrogenSamples will remain frozen for over 12 hoursRecyclable ethyl-vinyl (EVA) material offers a non-toxic alternative to conventional ice pans and bucketsDimensions (L x W x H): 40 x 31.5 x 12.5cm
Catalog No. NC2107331