Learn More
Medchemexpress LLC Laroprovstat (AZD0780) | 2455427-91-3 | 99.9% | 414.41 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14867350MG
Encompass_Preferred
Laroprovstat (AZD0780) is an orally active PCSK9 inhibitor. It has a binding affinity for PCSK9 with a Kd value of <200 nM. Laroprovstat binds to a pocket in the PCSK9 C-terminal domain without affecting the PCSK9-LDL receptor (LDLR) interaction. It inhibits the lysosomal trafficking of PSCK9-LDLR complexes and prevents PCSK9-induced LDLR degradation. It is used for the research of hypercholesterolemia.
  • Orally active PCSK9 inhibitor
  • High binding affinity for PCSK9 (Kd <200 nM)
  • Inhibits lysosomal trafficking of PSCK9-LDLR complexes
  • Prevents PCSK9-induced LDLR degradation
  • Potential for hypercholesterolemia research
Catalog No. 50-003-60043