Learn More
Medchemexpress LLC Larotinib mesylate hydrate | 2097129-93-4 | 98.7% | 699.17 g·mol⁻¹ | C26H36ClFN4O11S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY153110A10MG
Encompass_Preferred
Larotinib mesylate hydrate is an orally active, broad-spectrum tyrosine kinase inhibitor that primarily targets EGFR. Supplied as the mesylate hydrate salt for research use, it is provided as a solid with defined chemical and biological properties.
Catalog No. 50-002-85746