Learn More
Medchemexpress LLC Larotrectinib | 1223403-58-4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY12866
Encompass_Preferred
Larotrectinib (LOXO-101) is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors (TRKA, B, and C), showing low nanomolar 50% inhibitory concentrations and high selectivity. It demonstrates dose-dependent inhibition of cell proliferation, with good oral bioavailability and plasma protein binding in preclinical studies. This compound effectively reduces TRKA phosphorylation and downstream signaling, leading to tumor growth inhibition in vivo and reduced leukemic infiltration.
  • Selective inhibitor of TRK family receptors (TRKA, B, C)
  • Low nanomolar inhibitory concentrations and high selectivity
  • Dose-dependent inhibition of cell proliferation
  • Good oral bioavailability and plasma protein binding
  • Reduces TRKA phosphorylation and downstream signal transduction
  • Inhibits tumor growth in vivo
  • Reduces leukemic infiltration
Catalog No. 50-003-43050