Learn More
Medchemexpress LLC Larotrectinib | 1223403-58-4 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1286650MG
Encompass_Preferred
Larotrectinib (LOXO-101) is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors, demonstrating low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C). This product is intended for research and analytical applications.
  • Selective inhibitor of TRK family receptors
  • ATP-competitive mechanism of action
  • Inhibits TRKA, B, and C isoforms
  • Exhibits antiproliferative activity against various TRK fusion and mutant cell lines
  • For research use only
Catalog No. 50-003-41012