Learn More
TARGETMOL CHEMICALS INC LAROTRECTINIB 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T599550MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Larotrectinib (LOXO-101) is an orally administered inhibitor of the TRK kinase and is highly selective only for the TRK family of receptors(IC50s = 2-20 nM). purity: 100%
Catalog No. 50-257-9723