Learn More
Medchemexpress LLC Larotrectinib sulfate | 1223405-08-0 | 99.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12866A10MM/1ML
Encompass_Preferred
Larotrectinib sulfate is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors, with low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C).
  • ATP-competitive oral inhibitor
  • Selective inhibitor of TRK family receptors (TRKA, B, and C)
  • Low nanomolar 50% inhibitory concentrations against all three isoforms
  • Greater than 1,000-fold selectivity relative to other kinases
  • Demonstrates dose-dependent inhibition of cell proliferation
  • Reduces tyrosine phosphorylation of TRKA and downstream signal transduction
  • Well tolerated in GLP toxicology studies
  • Reduces leukemic infiltration to undetectable levels in bone marrow and spleen
Catalog No. 50-003-38246