Learn More
Medchemexpress LLC Larotrectinib sulfate | 1223405-08-0 | 99.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12866A10MG
Encompass_Preferred
Larotrectinib sulfate is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors, with low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C).
  • ATP-competitive oral inhibitor
  • Selective inhibitor of TRK family receptors
  • Low nanomolar inhibitory concentrations against TRKA, B, and C isoforms
Catalog No. 50-003-38223