Learn More
Selleck Chemical LLC Larotrectinib sulfate
SDP
Supplier:  Selleck Chemical LLC S796010MM/1ML
Encompass_Preferred
Larotrectinib sulfate is an oral potent and selective ATP-competitive inhibitor of tropomyosin receptor kinases (TRK) Larotrectinib inhibition of TRKs induces cellular apoptosis and G1 cell-cycle arrest
Catalog No. 50-000-00978