Learn More
Medchemexpress LLC Lartesertib | 2495096-26-7 | C23H21FN6O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1506171MG
Encompass_Preferred
Lartesertib (M4076) is a potent inhibitor of the serine/threonine protein kinase ATM. It has demonstrated the ability to inhibit the growth of various hematopoietic cell lines. When used in combination with the ATR inhibitor Tuvusertib (HY-111451), Lartesertib can induce tumor cell death, activate immune signaling pathways, and exhibit anti-tumor activity. It also alters the tumor microenvironment by upregulating PD-L1.
  • Potent inhibitor of ATM protein kinase
  • Inhibits growth of multiple hematopoietic cell lines
  • Induces tumor cell death when combined with Tuvusertib
  • Activates immune signaling pathways
  • Demonstrates anti-tumor activity
  • Upregulates PD-L1 on immune and tumor cells
  • Alters the tumor microenvironment
Catalog No. 50-003-75795