Learn More
Medchemexpress LLC Lartesertib | 2495096-26-7 | C23H21FN6O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15061710MM/1ML
Encompass_Preferred
Lartesertib, also known as M4076, is a potent inhibitor of the serine/threonine protein kinase ATM. It has been shown to inhibit the growth of multiple hematopoietic cell lines. When combined with the ATR inhibitor Tuvusertib, Lartesertib can promote the death of tumor cells, activate the immune signaling pathway, and exhibit significant anti-tumor activity.
  • Potent inhibitor of ATM protein kinase.
  • Inhibits growth of multiple hematopoietic cell lines.
  • Promotes tumor cell death in combination with Tuvusertib.
  • Activates the immune signaling pathway.
  • Exhibits anti-tumor activity.
Catalog No. 50-003-75154