Learn More
Medchemexpress LLC Lartesertib | 2495096-26-7 | C23H21FN6O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1506175MG
Encompass_Preferred
Lartesertib (M4076) is a highly potent inhibitor of the serine/threonine protein kinase ATM. It exhibits potential chemo-/radio-sensitizing and antineoplastic activities by inhibiting ATM kinase activity and signaling. This action prevents DNA damage checkpoint activation, disrupts DNA damage repair, and induces apoptosis in tumor cells.
  • Highly potent inhibitor of ATM kinase.
  • Inhibits the growth of various hematopoietic cell lines.
  • Promotes tumor cell death when combined with ATR inhibitor.
  • Activates the immune signaling pathway.
  • Exhibits anti-tumor activity.
  • Sensitizes tumor cells to chemo- and radiotherapy.
Catalog No. 50-003-75657