Learn More
AdipoGen Lasalocid A sodium salt, 25999-20-6, MFCD00078065, 250 mg
SDP
Supplier:  AdipoGen CDXL0015M250
Encompass_Preferred
MF: C34H53NaO8, MW: 612.8, ≥97%, Appearance: Colorless powder. Ionophore. Potent antibacterial agent and a coccidiostat. Able to form neutral complexes with monovalent and divalent cations and transport them through apolar phase (including lipid bilayer membranes). It can transport also larger organic cations, like protonated dopamine.
Catalog No. 50-117-7572